![Poloxamer liquid United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/320/746/253dda1a-6439-4d58-839b-4404693cf5f3/640/253dda1a-6439-4d58-839b-4404693cf5f3.png)
Poloxamer liquid
![Poloxamer liquid United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/320/746/253dda1a-6439-4d58-839b-4404693cf5f3/640/253dda1a-6439-4d58-839b-4404693cf5f3.png)
Synonym(s):
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)
CAS Number:
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing