Skip to Content
Merck
Pirimiphos-ethyl PESTANAL®, analytical standard

Pirimiphos-ethyl

Pirimiphos-ethyl PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C13H24N3O3PS
CAS Number:
Molecular Weight:
333.39
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing