Skip to Content
Merck
p-Fluorofentanyl solution 100 μg/mL in methanol, certified reference material, ampule of 0.5 mL, Cerilliant®

p-Fluorofentanyl solution

p-Fluorofentanyl solution 100 μg/mL in methanol, certified reference material, ampule of 0.5 mL, Cerilliant®
Empirical Formula (Hill Notation):
C22H27FN2O
CAS Number:
Molecular Weight:
354.46

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique