Skip to Content
Merck
Pantothenate for peak identification European Pharmacopoeia (EP) Reference Standard

Pantothenate for peak identification

Pantothenate for peak identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
D-Pantothenic acid hemicalcium salt, (R)-(+)-N-(2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl)-β-alanine hemicalcium salt, Calcium D-pantothenate, Vitamin B5
Linear Formula:
HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 ·1/2Ca
CAS Number:
Molecular Weight:
238.27

Products

Formula Weight
Manufacturer
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1