![Oxymetazoline Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/119/981/eef4e66c-d12c-4773-81f1-75fbd2270bc9/640/eef4e66c-d12c-4773-81f1-75fbd2270bc9.jpg)
Oxymetazoline Related Compound A
![Oxymetazoline Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/119/981/eef4e66c-d12c-4773-81f1-75fbd2270bc9/640/eef4e66c-d12c-4773-81f1-75fbd2270bc9.jpg)
Synonym(s):
N-(2-Aminoethyl)-2-[4-tert-butyl-3-hydroxy-2,6-dimethylphenyl]acetamide hydrochloride, N-(2-Aminoethyl)-2-(4-tert-butyl-3-hydroxy-2,6-dimethylphenyl)acetamide hydrochloride
Empirical Formula (Hill Notation):
C16H26N2O2·HCl
CAS Number:
Molecular Weight:
314.85
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2