
Oridonin

Synonym(s):
7a,20-Epoxy-1a,6b,7,14-tetrahydroxy-Kaur-16-en-15-one, Isodonol
Empirical Formula (Hill Notation):
C20H28O6
CAS Number:
Molecular Weight:
364.43
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing