![ORBO™ 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50 W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea](/deepweb/assets/sigmaaldrich/product/images/175/546/5155e6d4-ffb9-4f3e-8615-fbcb94a1fa51/640/5155e6d4-ffb9-4f3e-8615-fbcb94a1fa51.jpg)
ORBO™ 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50
![ORBO™ 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50 W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea](/deepweb/assets/sigmaaldrich/product/images/175/546/5155e6d4-ffb9-4f3e-8615-fbcb94a1fa51/640/5155e6d4-ffb9-4f3e-8615-fbcb94a1fa51.jpg)
Synonym(s):
ORBO™ Activated Silica Gel Tube
Products
Brand
Product Line
Agency
Chemical Composition
OD (mm)
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing