
Nitrosotriaminopyrimidine

Synonym(s):
2,4,6-Triamino-5-nitrosopyrimidine
Empirical Formula (Hill Notation):
C4H6N6O
CAS Number:
Molecular Weight:
154.13
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing