![Niflumic acid impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/162/529/8340794e-3d9a-4ad4-8d8e-fd16ac3140fc/640/8340794e-3d9a-4ad4-8d8e-fd16ac3140fc.png)
Niflumic acid impurity A
![Niflumic acid impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/162/529/8340794e-3d9a-4ad4-8d8e-fd16ac3140fc/640/8340794e-3d9a-4ad4-8d8e-fd16ac3140fc.png)
Synonym(s):
2-Chloropyridine-3-carboxylic acid, 2-Chloronicotinic acid
Empirical Formula (Hill Notation):
C6H4ClNO2
CAS Number:
Molecular Weight:
157.55
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing