Skip to Content
Merck
NIDA-41020 ≥97% (HPLC), solid

NIDA-41020

NIDA-41020 ≥97% (HPLC), solid
Synonym(s):
1-(2,4-Dichlorophenyl)-5-(4-methoxyphenyl)-4-methyl-N-(1-piperidinyl)-1H-pyrazole-3-carboxamide
Empirical Formula (Hill Notation):
C23H24Cl2N4O2
CAS Number:
Molecular Weight:
459.37

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing