Skip to Content
Merck
N-Ethyl-D-glucamine ≥98.0%

N-Ethyl-D-glucamine

N-Ethyl-D-glucamine ≥98.0%
Empirical Formula (Hill Notation):
C8H19NO5
CAS Number:
Molecular Weight:
209.24
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing