Skip to Content
Merck
N-4-Boc-aminocyclohexanone 95%

N-4-Boc-aminocyclohexanone

N-4-Boc-aminocyclohexanone 95%
Synonym(s):
(4-Oxocyclohexyl)carbamicacid tert-butyl ester, 4-(tert-Butoxycarbonylamino)cyclohexanone, tert-Butyl 4-oxocyclohexylcarbamate
Empirical Formula (Hill Notation):
C11H19NO3
CAS Number:
Molecular Weight:
213.27

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing