Skip to Content
Merck
Morphine-D3 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Morphine-D3 solution

Morphine-D3 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C17H16NO3D3
CAS Number:
Molecular Weight:
288.36

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique