
Monocrotaline N-oxide

Empirical Formula (Hill Notation):
C16H23NO7
CAS Number:
Molecular Weight:
341.36
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing