
Monoclonal Anti-CD3-PE antibody produced in mouse

Synonym(s):
Anti-CD3 (activation epitope)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2