
Monacolin K

Synonym(s):
Lovastatin, Mevinolin
Empirical Formula (Hill Notation):
C24H36O5
CAS Number:
Molecular Weight:
404.54
Products
Formula Weight
Manufacturer
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing