![Midazolam hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/395/534/24f8f4c5-0bc4-4965-b32d-a91c0f216f7b/640/24f8f4c5-0bc4-4965-b32d-a91c0f216f7b.png)
Midazolam hydrochloride
![Midazolam hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/395/534/24f8f4c5-0bc4-4965-b32d-a91c0f216f7b/640/24f8f4c5-0bc4-4965-b32d-a91c0f216f7b.png)
Synonym(s):
8-Chloro-6-(2-fluorophenyl)-1-methyl-4H-imidazo[1,5a][1,4]benzodiazepine hydrochloride
Empirical Formula (Hill Notation):
C18H13ClFN3 · HCl
CAS Number:
Molecular Weight:
362.23
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing