![Metronidazole Impurity G certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/366/963/d00b28e4-6c0c-464c-b6eb-6eea28f000cd/640/d00b28e4-6c0c-464c-b6eb-6eea28f000cd.png)
Metronidazole Impurity G
![Metronidazole Impurity G certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/366/963/d00b28e4-6c0c-464c-b6eb-6eea28f000cd/640/d00b28e4-6c0c-464c-b6eb-6eea28f000cd.png)
Synonym(s):
2-(2-Methyl-5-nitro-1H-imidazol-1-yl)acetic acid, 1-Carboxymethylmetronidazole, 2-Methyl-5-nitroimidazole-1-acetic acid, Metronidazole-1-acetic acid
Empirical Formula (Hill Notation):
C6H7N3O4
CAS Number:
Molecular Weight:
185.14
Products
Brand
Formula Weight
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing