![Metronidazole impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/375/336/e2eed2dd-612a-459a-897d-4b032dbf99e0/640/e2eed2dd-612a-459a-897d-4b032dbf99e0.png)
Metronidazole impurity A
![Metronidazole impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/375/336/e2eed2dd-612a-459a-897d-4b032dbf99e0/640/e2eed2dd-612a-459a-897d-4b032dbf99e0.png)
Synonym(s):
2-Methyl-4(5)-nitroimidazole
Empirical Formula (Hill Notation):
C4H5N3O2
CAS Number:
Molecular Weight:
127.10
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing