![Methylergometrine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/311/636/89c62d08-8a44-40d3-9d5a-3206da116efa/640/89c62d08-8a44-40d3-9d5a-3206da116efa.png)
Methylergometrine for system suitability
![Methylergometrine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/311/636/89c62d08-8a44-40d3-9d5a-3206da116efa/640/89c62d08-8a44-40d3-9d5a-3206da116efa.png)
Synonym(s):
Methylergonovine maleate salt, 9,10-Didehydro-N-[(S)-1-(hydroxymethyl)propyl]-6-methylergoline-8β-carboxamide maleate salt, Methergine maleate salt
Empirical Formula (Hill Notation):
C20H25N3O2 · C4H4O4
CAS Number:
Molecular Weight:
455.50
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing