Methyl blue (C.I. 42780)
Empirical Formula (Hill Notation):
C37H27N3Na2O9S3
CAS Number:
Molecular Weight:
799.80
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing