
Methyl 2-chloropropionate

Synonym(s):
Methyl 2-chloropropionate
Empirical Formula (Hill Notation):
C4H7ClO2
CAS Number:
Molecular Weight:
122.55
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing