Skip to Content
Merck
Methotrexate hydrate ≥99.0% (sum of enantiomers, HPLC)

Methotrexate hydrate

Methotrexate hydrate ≥99.0% (sum of enantiomers, HPLC)
Synonym(s):
4-Amino-10-methylfolic acid hydrate, L-4-Amino-N10-methylpteroylglutamic acid hydrate, L-Amethopterin hydrate, Antifolan hydrate, MTX hydrate, Methylaminopterin hydrate
Empirical Formula (Hill Notation):
C20H22N8O5 · xH2O
CAS Number:
Molecular Weight:
454.44 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Activity Spectrum
Quality Segment
Markush Class
Markush Group
Mode of Action
Technique