![Meperfluthrin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/343/696/c52936a8-8d71-4820-85e2-e306cbeab23f/640/c52936a8-8d71-4820-85e2-e306cbeab23f.png)
Meperfluthrin
![Meperfluthrin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/343/696/c52936a8-8d71-4820-85e2-e306cbeab23f/640/c52936a8-8d71-4820-85e2-e306cbeab23f.png)
Synonym(s):
2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl (1R,3S)-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate
Empirical Formula (Hill Notation):
C17H16Cl2F4O3
CAS Number:
Molecular Weight:
415.21
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing