Skip to Content
Merck
m-Cymene analytical standard

m-Cymene

m-Cymene analytical standard
Synonym(s):
1-Isopropyl-3-methylbenzene, 3-Isopropyltoluene
Linear Formula:
CH3C6H4CH(CH3)2
CAS Number:
Molecular Weight:
134.22
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing