Skip to Content
Merck
m-Chlorobenzoic acid NIST® SRM® 2144

m-Chlorobenzoic acid

m-Chlorobenzoic acid NIST® SRM® 2144
Synonym(s):
3-Chlorobenzoic acid
Linear Formula:
ClC6H4CO2H
CAS Number:
Molecular Weight:
156.57

Products

Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing