Skip to Content
Merck
L-Phenylalanine-1-13C endotoxin tested, 99 atom % 13C, 99% (CP)

L-Phenylalanine-1-13C

L-Phenylalanine-1-13C endotoxin tested, 99 atom % 13C, 99% (CP)
Synonym(s):
L-Phenylalanine-carboxy-13C
Linear Formula:
C6H5CH2CH(NH2)13CO2H
CAS Number:
Molecular Weight:
166.18

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing