
Lorazepam Related Compound B

Synonym(s):
2-Amino-2′,5-dichlorobenzophenone
Linear Formula:
H2NC6H3(Cl)COC6H4Cl
CAS Number:
Molecular Weight:
266.12
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing