Skip to Content
Merck
L-(−)-Fucose analytical standard

L-(−)-Fucose

L-(−)-Fucose analytical standard
Synonym(s):
6-Deoxy-L-galactose, L-(−)-Fucose
Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing