Skip to Content
Merck
Kurarinone ≥98% (HPLC)

Kurarinone

Kurarinone ≥98% (HPLC)
Synonym(s):
(-)-Kurarinone, (2S)-2-(2,4-Dihydroxyphenyl)-2,3-dihydro-7-hydroxy-5-methoxy-8-[(2R)-5-methyl-2-(1-methylethenyl)-4-hexen-1-yl]-4H-1-benzopyran-4-one, Marini
Empirical Formula (Hill Notation):
C26H30O6
CAS Number:
Molecular Weight:
438.51

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing