Skip to Content
Merck
Ketoisoleucine sodium salt certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Ketoisoleucine sodium salt

Ketoisoleucine sodium salt certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(±)-3-Methyl-2-oxovaleric acid sodium salt, (±)-Sodium 3-methyl-2-oxovalerate, 3-Methyl-2-oxopentanoic acid sodium salt, DL-α-Keto-β-methylvaleric acid sodium salt, Ketoisoleucine sodium salt
Linear Formula:
CH3CH2CH(CH3)COCOONa
CAS Number:
Molecular Weight:
152.12

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Reference Material Type