
Isradipine Related Compound A

Synonym(s):
Methyl 1-methylethyl 4-(2,1,3-benzoxadiazol-4-yl) -2,6-dimethylpyridine-3,5-dicarboxylate, Isopropyl methyl 4-(4-benzofurazanyl)-2,6-dimethyl-3,5-pyridinedicarboxylate
Empirical Formula (Hill Notation):
C19H19N3O5
CAS Number:
Molecular Weight:
369.37
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing