Isomaltotriose, DP3
Synonym(s):
Isomaltotriose, α-D-Glucosyl-[1→6]-α-D-glucosyl-[1→6]-D-glucose
Empirical Formula (Hill Notation):
C18H32O16
CAS Number:
Molecular Weight:
504.44
Products
Brand
Formula Weight
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing