[Ir(dtbbpy)(ppy)2]PF6
Synonym(s):
[Ir(dtbbpy)(ppy)2][PF6]
Empirical Formula (Hill Notation):
C40H40F6IrN4P
CAS Number:
Molecular Weight:
913.95
Products
Brand
Formula Weight
Physical Form
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing