Skip to Content
Merck
Idaein chloride ≥90% (HPLC)

Idaein chloride

Idaein chloride ≥90% (HPLC)
Synonym(s):
3-(Galactosyloxy)-3′,4′,5,7-tetrahydroxyflavylium chloride, Cyanidin 3-O-β-D-galactopyranoside, Cyanidin 3-O-galactoside chloride, Idein chloride
Empirical Formula (Hill Notation):
C21H21ClO11
CAS Number:
Molecular Weight:
484.84

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing