Guanidine Hydrochloride
Synonym(s):
Guanidinium chloride
Empirical Formula (Hill Notation):
CH5N3 · xHCl
CAS Number:
Molecular Weight:
59.07 (free base basis)
Products
Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing