Skip to Content
HK
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Graphene ink
Synonym(s):
conductive ink
Products
Brand
Sigma-Aldrich (3)
Sustainability
Boiling Point (°C)
Feature
Physical Form
Chemical Composition
Quality Segment
Particle Size (µm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
793663
Graphene ink,
for inkjet printing, with ethyl cellulose in cyclohexanone and terpineol, inkjet printable
Expand
View Pricing
900695
Graphene ink,
for inkjet printing, photonically annealable
Expand
View Pricing
796115
Graphene ink,
for gravure printing, with ethyl cellulose in terpineol, gravure printable
Expand
View Pricing
798983
Graphene ink,
for screen printing, with ethyl cellulose in terpineol, screen printable
Expand
View Pricing