Granisetron Related Compound C
Synonym(s):
N-[(1R,3r,5S)-9-Azabicyclo[3.3.1]non-3-yl]-1-methyl-1H-indazole-3-carboxamide monohydrochloride, N-Demethyl-endo-granisetron hydrochloride, endo-N-9-Azabicyclo[3.3.1]non-3-yl-1-methyl-1H-indazole-3-carboxamide monohydrochloride
Empirical Formula (Hill Notation):
C17H22N4O · HCl
CAS Number:
Molecular Weight:
334.84
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing