
Glycerol monolinoleate

Empirical Formula (Hill Notation):
C21H38O4
CAS Number:
Molecular Weight:
354.52
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing