![Geranium thiazole ≥98%, FG](/deepweb/assets/sigmaaldrich/product/structures/102/176/0237d0e8-4ac3-4cf6-9ee3-f7091a7681c9/640/0237d0e8-4ac3-4cf6-9ee3-f7091a7681c9.png)
Geranium thiazole
![Geranium thiazole ≥98%, FG](/deepweb/assets/sigmaaldrich/product/structures/102/176/0237d0e8-4ac3-4cf6-9ee3-f7091a7681c9/640/0237d0e8-4ac3-4cf6-9ee3-f7091a7681c9.png)
Synonym(s):
4,5-Dimethyl-2-isobutyl thiazole
Empirical Formula (Hill Notation):
C9H15NS
CAS Number:
Molecular Weight:
169.29
Products
Product Category
Brand
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing