Skip to Content
Merck
vibrant-m

Fuchsin acid (C.I. 42685)

vibrant-m
Empirical Formula (Hill Notation):
C20H17N3Na2O9S3
CAS Number:
Molecular Weight:
585.54

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing