Skip to Content
Merck
Foscarnet Related Compound B pharmaceutical secondary standard, certified reference material

Foscarnet Related Compound B

Foscarnet Related Compound B pharmaceutical secondary standard, certified reference material
Synonym(s):
1-Ethoxy-1-hydroxyphosphinecarboxylic acid 1-oxide disodium salt, Disodium (ethoxyoxydophosphanyl)formate
Empirical Formula (Hill Notation):
C3H5Na2O5P
CAS Number:
Molecular Weight:
198.02

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade