Skip to Content
Merck
vibrant-m

Fluticasone furoate for system suitability

vibrant-m
Synonym(s):
Fluticasone furoate, 6α,9α-Difluoro-17α-[(2-furanylcarbonyl)oxy]-11β-hydroxy-6α-methyl-3-oxoandrosta-1,4-diene-17β-carbothioic acid S-fluoromethyl ester
Empirical Formula (Hill Notation):
C27H29F3O6S
CAS Number:
Molecular Weight:
538.58

Products

Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Showing 1-1 of 1