
Fluosilicic acid

Synonym(s):
Fluorosilicic acid, Hexafluorosilicic acid, Hydrogen hexafluorosilicate
Linear Formula:
H2SiF6
CAS Number:
Molecular Weight:
144.09
Products
Product Category
Brand
Formula Weight
Quality Segment
Special Grade
Showing 1-1 of 1