![Fipronil for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/374/889/9543861a-f2ec-4daf-b6b2-1df502a207cb/640/9543861a-f2ec-4daf-b6b2-1df502a207cb.png)
Fipronil for system suitability
![Fipronil for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/374/889/9543861a-f2ec-4daf-b6b2-1df502a207cb/640/9543861a-f2ec-4daf-b6b2-1df502a207cb.png)
Synonym(s):
Fipronil
Empirical Formula (Hill Notation):
C12H4Cl2F6N4OS
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing