Skip to Content
Merck
Ethylenediaminetetraacetic acid disodium magnesium salt tetrahydrate SAJ special grade, ≥99.0%

Ethylenediaminetetraacetic acid disodium magnesium salt tetrahydrate

Ethylenediaminetetraacetic acid disodium magnesium salt tetrahydrate SAJ special grade, ≥99.0%
Synonym(s):
Ethylenediaminetetraacetic acid disodium magnesium salt
Linear Formula:
(NaOOCCH2)2NCH2CH2N(CH2COO)2Mg · 4H2O
CAS Number:
Molecular Weight:
430.56
EC Number:

Products

Brand
Formula Weight
Purity
Showing 1-1 of 1