Ethyl Acetate Biosynthesis
Synonym(s):
Ethyl acetate, EtOAc
Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing