Skip to Content
Merck
Ethyl 4-hydroxy-2-methyl-1,1-dioxo-2H-1,2-benzothiazine-3-carboxylate 97%

Ethyl 4-hydroxy-2-methyl-1,1-dioxo-2H-1,2-benzothiazine-3-carboxylate

Ethyl 4-hydroxy-2-methyl-1,1-dioxo-2H-1,2-benzothiazine-3-carboxylate 97%
Empirical Formula (Hill Notation):
C12H13NO5S
CAS Number:
Molecular Weight:
283.30
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing