Eschka’s Reagent
Synonym(s):
Magnesium oxide – sodium carbonate 2:1
Linear Formula:
2MgO · Na2CO3
CAS Number:
Products
Brand
Physical Form
Purity
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing