![Disialoganglioside GD1a from bovine brain ≥95% (TLC), lyophilized powder](/deepweb/assets/sigmaaldrich/product/structures/209/819/e0c0664b-0d66-4823-9edd-3438efb8f05d/640/e0c0664b-0d66-4823-9edd-3438efb8f05d.png)
Disialoganglioside GD1a from bovine brain
![Disialoganglioside GD1a from bovine brain ≥95% (TLC), lyophilized powder](/deepweb/assets/sigmaaldrich/product/structures/209/819/e0c0664b-0d66-4823-9edd-3438efb8f05d/640/e0c0664b-0d66-4823-9edd-3438efb8f05d.png)
Synonym(s):
Ganglioside GD1a, disialo from bovine brain
CAS Number:
Products
Product Category
Brand
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing